ENAH31552171 Enamine

1-phenyl-3-(pyridin-2-yl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD31746239
  • Cas Number : 2166046-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739393
Availability details for
Quantity
  • Availability
  • Add to Cart