ENAH31552173 Enamine

1-(cyclopropylamino)-3,3-dimethylbutan-2-ol

  • MDL number MFCD17218798
  • Cas Number : 1310305-98-6
  • Molecular Weight : 157.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739409
Availability details for
Quantity
  • Availability
  • Add to Cart