ENAH31552174 Enamine

5-methyl-2-sulfamoylthiophene-3-carboxylic acid

  • MDL number MFCD31746240
  • Cas Number : 2287274-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739410
Availability details for
Quantity
  • Availability
  • Add to Cart