ENAH31552177 Enamine

3-(azetidin-3-yl)-4,5-dihydro-1,2-oxazole; trifluoroacetic acid

  • MDL number MFCD31746471

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739420
Availability details for
Quantity
  • Availability
  • Add to Cart