ENAH31552179 Enamine

3-(pyrrolidin-3-yl)-4,5-dihydro-1,2-oxazole; trifluoroacetic acid

  • MDL number MFCD31746243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739443
Availability details for
Quantity
  • Availability
  • Add to Cart