ENAH3155217A Enamine

tert-butyl (2R)-4-(carboxy)-2-methylpiperazine-1-carboxylate

  • MDL number MFCD31746472
  • Cas Number : 2287238-45-1
  • Molecular Weight : 262.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739463
Availability details for
Quantity
  • Availability
  • Add to Cart