ENAH3155217B Enamine

ethyl 2-iodo-1,3-oxazole-5-carboxylate

  • MDL number MFCD28137528
  • Cas Number : 220580-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6735348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735348
Availability details for
Quantity
  • Availability
  • Add to Cart