ENAH3155217D Enamine

methyl 1-(hydroxymethyl)cyclopent-3-ene-1-carboxylate

  • MDL number MFCD31746246
  • Cas Number : 2287286-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739481
Availability details for
Quantity
  • Availability
  • Add to Cart