ENAH31552188 Enamine

tert-butyl N-{7-aminospiro[3.5]nonan-2-yl}carbamate hydrochloride

  • MDL number MFCD31746300

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739568
Availability details for
Quantity
  • Availability
  • Add to Cart