ENAH3155218A Enamine

2,3-bis(benzyloxy)propan-1-ol

  • MDL number MFCD00792598
  • Cas Number : 59991-89-8
  • Molecular Weight : 272.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6739578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739578
Availability details for
Quantity
  • Availability
  • Add to Cart