ENAH3155218B Enamine

2-(piperidin-1-yl)-2-(pyridin-3-yl)acetamide

  • MDL number MFCD19318161
  • Cas Number : 856858-17-8
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739579
Availability details for
Quantity
  • Availability
  • Add to Cart