ENAH31552192 Enamine

3-bromo-5-(dimethylphosphoryl)aniline

  • MDL number MFCD31746305
  • Cas Number : 2287315-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739604
Availability details for
Quantity
  • Availability
  • Add to Cart