ENAH31552193 Enamine

sodium 5-methoxy-1,3-dimethyl-1H-pyrazole-4-sulfinate

  • MDL number MFCD31746477
  • Cas Number : 2287311-21-9
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739605
Availability details for
Quantity
  • Availability
  • Add to Cart