ENAH31552198 Enamine

tert-butyl 4-[(chlorocarbonyl)(methyl)amino]piperidine-1-carboxylate

  • MDL number MFCD30294662
  • Cas Number : 1383546-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739642
Availability details for
Quantity
  • Availability
  • Add to Cart