ENAH31552199 Enamine

tert-butyl 4-(carboxy)-3-methylpiperazine-1-carboxylate

  • MDL number MFCD31794895
  • Cas Number : 1626387-98-1
  • Molecular Weight : 262.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739643
Availability details for
Quantity
  • Availability
  • Add to Cart