ENAH3155219B Enamine

tert-butyl (2R,6S)-4-(carboxy)-2,6-dimethylpiperazine-1-carboxylate

  • MDL number MFCD31794896
  • Cas Number : 1098598-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739654
Availability details for
Quantity
  • Availability
  • Add to Cart