ENAH315521A2 Enamine

10-oxa-6-azaspiro[4.6]undecane hydrochloride

  • MDL number MFCD31746480
  • Cas Number : 2287280-15-1
  • Molecular Weight : 191.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739697
Availability details for
Quantity
  • Availability
  • Add to Cart