ENAH315521B3 Enamine

3-[(tert-butoxy)methyl]piperidine hydrochloride

  • MDL number MFCD31746485
  • Cas Number : 1799284-77-7
  • Molecular Weight : 207.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739761
Availability details for
Quantity
  • Availability
  • Add to Cart