ENAH315521BF Enamine

4-methyl-5H,6H,7H-pyrrolo[3,4-b]pyridin-7-one

  • MDL number MFCD31794917
  • Cas Number : 2287340-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739877
Availability details for
Quantity
  • Availability
  • Add to Cart