ENAH315521C5 Enamine

5-bromo-1-cyclopropyl-6-methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid

  • MDL number MFCD31746516
  • Cas Number : 1569649-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739932
Availability details for
Quantity
  • Availability
  • Add to Cart