ENAH315521C6 Enamine

1-(2-hydroxyethyl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD10005852
  • Cas Number : 1176646-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739934
Availability details for
Quantity
  • Availability
  • Add to Cart