ENAH315521C8 Enamine

(2R)-1-cyclohexylpropan-2-amine hydrochloride

  • MDL number MFCD31800452
  • Cas Number : 56401-85-5
  • Molecular Weight : 177.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739948
Availability details for
Quantity
  • Availability
  • Add to Cart