ENAH315521CD Enamine

2,2,2-trifluoro-1-(5,6,7,8-tetrahydropteridin-5-yl)ethan-1-one

  • MDL number MFCD31737142
  • Cas Number : 2260936-53-4
  • Molecular Weight : 232.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6735480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735480
Availability details for
Quantity
  • Availability
  • Add to Cart