ENAH315521D3 Enamine

tert-butyl 1-methyl-1,5,6,7-tetrahydrospiro[imidazo[4,5-c]pyridine-4,3'-pyrrolidine]-1'-carboxylate

  • MDL number MFCD31746522
  • Cas Number : 2287332-99-2
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740008
Availability details for
Quantity
  • Availability
  • Add to Cart