ENAH315521D9 Enamine

[3-(chloromethyl)phenyl](imino)propyl-lambda6-sulfanone hydrochloride

  • MDL number MFCD31746558
  • Cas Number : 2287288-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740034
Availability details for
Quantity
  • Availability
  • Add to Cart