ENAH315521EC Enamine

3-[4-(2-methoxyphenyl)piperazin-1-yl]propane-1,2-diol

  • MDL number MFCD05269408
  • Cas Number : 117067-06-8
  • Molecular Weight : 266.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740138
Availability details for
Quantity
  • Availability
  • Add to Cart