ENAH315521F6 Enamine

(1r,3r)-3-(ethoxycarbonyl)-1-fluorocyclobutane-1-carboxylic acid

  • MDL number MFCD31746593
  • Cas Number : 2287280-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740194
Availability details for
Quantity
  • Availability
  • Add to Cart