ENAH315521F7 Enamine

(1s,3s)-3-(ethoxycarbonyl)-1-fluorocyclobutane-1-carboxylic acid

  • Cas Number : 2287298-76-2
  • Molecular Weight : 190.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6740196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740196
Availability details for
Quantity
  • Availability
  • Add to Cart