ENAH315521F8 Enamine

(4R)-4-(propan-2-yl)-1lambda6,2,5-thiadiazolidine-1,1,3-trione

  • MDL number MFCD31746568
  • Cas Number : 2287238-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740209
Availability details for
Quantity
  • Availability
  • Add to Cart