ENAH31552200 Enamine

6-amino-1-({[(tert-butoxy)carbonyl]amino}amino)-2,2-dimethylhexan-1-one

  • MDL number MFCD29037696
  • Cas Number : 2034155-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740252
Availability details for
Quantity
  • Availability
  • Add to Cart