ENAH31552205 Enamine

2-(chloromethyl)-4-(trifluoromethyl)thiophene

  • MDL number MFCD31746601
  • Cas Number : 1643965-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740295

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6740295
pictograms GHS05,GHS07
hstatements H227,H302,H312,H314,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart