ENAH3155220A Enamine

N,1-dimethyl-2,6,7-trioxabicyclo[2.2.2]octan-4-amine

  • MDL number MFCD31746573
  • Cas Number : 2287314-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740313

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740313
Availability details for
Quantity
  • Availability
  • Add to Cart