ENAH31552216 Enamine

tert-butyl 4-[(methylamino)methyl]-2,3-dihydro-1H-pyrrole-1-carboxylate hydrochloride

  • MDL number MFCD31746664
  • Molecular Weight : 248.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740334
Availability details for
Quantity
  • Availability
  • Add to Cart