ENAH31552224 Enamine

4-cyclobutyl-1,2-oxazol-5-amine

  • MDL number MFCD31794925
  • Cas Number : 1779920-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740441
Availability details for
Quantity
  • Availability
  • Add to Cart