ENAH31552226 Enamine

3-(chloromethyl)-5,5-dimethyl-4,5-dihydro-1,2-oxazole

  • MDL number MFCD19236134
  • Cas Number : 1314981-14-0
  • Molecular Weight : 147.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740464
Availability details for
Quantity
  • Availability
  • Add to Cart