ENAH3155222D Enamine

1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-1,2,3,6-tetrahydropyridine-3-carboxylic acid

  • MDL number MFCD02682420
  • Cas Number : 1262769-81-2
  • Molecular Weight : 349.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740483
Availability details for
Quantity
  • Availability
  • Add to Cart