ENAH3155223C Enamine

5-(dimethylphosphoryl)-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD31794916
  • Cas Number : 2254055-98-4
  • Molecular Weight : 209.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740573
Availability details for
Quantity
  • Availability
  • Add to Cart