ENAH3155224C Enamine

4-(trifluoromethyl)-1H,2H,5H,6H,7H,8H-pyrido[3,4-d]pyridazin-1-one hydrochloride

  • MDL number MFCD31794594
  • Cas Number : 2287274-40-0
  • Molecular Weight : 255.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6740726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740726
Availability details for
Quantity
  • Availability
  • Add to Cart