ENAH31552250 Enamine

1-methyl-5-(3,3,3-trifluoropropyl)-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD31794595
  • Cas Number : 2287322-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740747
Availability details for
Quantity
  • Availability
  • Add to Cart