ENAH31552252 Enamine

3-isocyanato-5-methoxy-1H-indole

  • MDL number MFCD31794596

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740771

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P310,P302+P352,P304+P340,P304+P341,P305+P351+P338,P311,P312,P330,P332+P313,P337+P313,P342+P311,P361,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6740771
pictograms GHS06,GHS08
hstatements H301,H311,H315,H319,H331,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart