ENAH31552253 Enamine

5-(3-iodophenyl)-1-methyl-1H-1,2,4-triazole

  • MDL number MFCD31794533
  • Cas Number : 2287317-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740772
Availability details for
Quantity
  • Availability
  • Add to Cart