ENAH3155225D Enamine

2-cyano-N-(2,3-dichlorophenyl)-2-methylacetamide

  • MDL number MFCD16121210
  • Cas Number : 1251387-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740864
Availability details for
Quantity
  • Availability
  • Add to Cart