ENAH3155225E Enamine

2,2,2-trifluoro-1-(5-fluoro-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD15524799
  • Cas Number : 1260793-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740884
Availability details for
Quantity
  • Availability
  • Add to Cart