ENAH31552265 Enamine

tert-butyl 2-isothiocyanatoacetate

  • MDL number MFCD28355505
  • Cas Number : 58511-02-7
  • Molecular Weight : 173.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740912
Availability details for
Quantity
  • Availability
  • Add to Cart