ENAH3155226B Enamine

(2E)-N-(5-bromo-2-chlorophenyl)-3-phenylprop-2-enamide

  • MDL number MFCD18039100

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740935
Availability details for
Quantity
  • Availability
  • Add to Cart