ENAH3155226D Enamine

ethyl 3-(1H-1,2,4-triazol-5-yl)propanoate hydrochloride

  • MDL number MFCD31794631
  • Cas Number : 2287310-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740957
Availability details for
Quantity
  • Availability
  • Add to Cart