ENAH31552276 Enamine

1-{1H-pyrrolo[2,3-d]pyrimidin-4-yl}piperazine dihydrochloride

  • MDL number MFCD21604134
  • Cas Number : 853680-06-5
  • Molecular Weight : 276.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6741056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6741056
Availability details for
Quantity
  • Availability
  • Add to Cart