ENAH31552279 Enamine

5-ethenyl-2,3-dimethylpyridine

  • MDL number MFCD18256339
  • Cas Number : 113989-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6742990

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6742990
pictograms GHS05,GHS07
hstatements H227,H302,H312,H314,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart