ENAH3155230A Enamine

3-chloro-5-methylpyridin-1-ium-1-olate

  • MDL number MFCD31794816
  • Cas Number : 97944-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743442
Availability details for
Quantity
  • Availability
  • Add to Cart