ENAH31552313 Enamine

methyl 2-amino-3-(6-chloropyridin-3-yl)propanoate hydrochloride

  • MDL number MFCD31794835
  • Cas Number : 2287259-57-6
  • Molecular Weight : 251.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743909
Availability details for
Quantity
  • Availability
  • Add to Cart